Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046500.2 | NP_001039965.1 | 2 - 3 | 433 | 113 | |
GenBank mRNA | BC110266.1 | - | 2 - 3 | 435 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |