Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046502.2 | NP_001039967.1 | 9 - 10 | 1761 | 68 | |
GenBank mRNA | BC105377.1 | - | 9 - 10 | 1763 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |