Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC22C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046546.2 | NP_001040011.1 | 3 - 4 | 539 | 107 | |
GenBank mRNA | BC110195.1 | - | 3 - 4 | 540 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |