Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075220.2 | NP_001068688.2 | 1 - 2 | 285 | 110 | |
GenBank mRNA | BC110154.1 | - | 1 - 2 | 286 | 110 | |
DT856624.1 | - | 1 - 2 | 286 | 110 | ||
EE348237.1 | - | 1 - 2 | 284 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |