Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075377.2 | NP_001068845.1 | 2 - 3 | 395 | 69 | |
GenBank mRNA | BC111657.1 | - | 2 - 3 | 396 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |