Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075866.2 | NP_001069334.1 | 6 - 7 | 845 | 68 | |
GenBank mRNA | BC112612.1 | - | 6 - 7 | 846 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |