Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076094.1 | NP_001069562.1 | 4 - 5 | 1102 | 105 | |
GenBank mRNA | BC109887.1 | - | 4 - 5 | 1102 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |