Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076240.2 | NP_001069708.1 | 3 - 4 | 577 | 81 | |
GenBank mRNA | BC109997.1 | - | 3 - 4 | 577 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |