Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3H1orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076346.1 | NP_001069814.1 | 1 - 2 | 73 | 103 | |
GenBank mRNA | BC118464.1 | - | 1 - 2 | 73 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |