Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076402.2 | NP_001069870.1 | 3 - 4 | 325 | 61 | |
GenBank mRNA | BC109788.1 | - | 3 - 4 | 326 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |