Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076402.2 | NP_001069870.1 | 7 - 8 | 755 | 147 | |
GenBank mRNA | BC109788.1 | - | 7 - 8 | 756 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |