Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076408.2 | NP_001069876.1 | 3 - 4 | 431 | 82 | |
GenBank mRNA | BC111146.1 | - | 3 - 4 | 432 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |