Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNIP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076935.2 | NP_001070403.1 | 8 - 9 | 768 | 144 | |
GenBank mRNA | BC112750.1 | - | 8 - 9 | 769 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |