Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASRGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077035.2 | NP_001070503.1 | 3 - 4 | 461 | 64 | |
GenBank mRNA | BC109621.1 | - | 3 - 4 | 462 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |