Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5H12orf29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077047.2 | NP_001070515.1 | 6 - 7 | 948 | 99 | |
GenBank mRNA | BC105491.1 | - | 6 - 7 | 949 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |