Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077052.2 | NP_001070520.1 | 4 - 5 | 408 | 125 | |
GenBank mRNA | BC109640.1 | - | 4 - 5 | 409 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |