Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C29H11orf86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077090.2 | NP_001070558.1 | 1 - 2 | 323 | 83 | |
GenBank mRNA | BC112590.1 | - | 1 - 2 | 324 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |