Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077094.2 | NP_001070562.1 | 12 - 13 | 1314 | 90 | |
GenBank mRNA | BC112778.1 | - | 12 - 13 | 1316 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |