Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSGIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077129.2 | NP_001070597.1 | 4 - 5 | 552 | 147 | |
GenBank mRNA | BC112571.1 | - | 4 - 5 | 553 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |