Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078060.1 | NP_001071528.1 | 3 - 4 | 910 | 60 | |
GenBank mRNA | BC109527.1 | - | 3 - 4 | 910 | 60 | |
FJ790227.1 | - | 3 - 4 | 773 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |