Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078064.2 | NP_001071532.1 | 196 | 122 | ||
GenBank mRNA | BC111656.1 | - | 197 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus |