Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079619.2 | NP_001073087.1 | 2 - 3 | 123 | 112 | |
GenBank mRNA | BC112592.1 | - | 2 - 3 | 124 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |