Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079644.2 | NP_001073112.1 | 1 - 2 | 827 | 139 | |
GenBank mRNA | BC111293.1 | - | 1 - 2 | 828 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |