Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTI1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079647.2 | NP_001073115.1 | 1 - 2 | 250 | 110 | |
GenBank mRNA | BC112511.1 | - | 1 - 2 | 251 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |