Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079799.1 | NP_001073267.1 | 5 - 6 | 677 | 79 | |
GenBank mRNA | BC112534.1 | - | 5 - 6 | 677 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |