Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015627.2 | NP_001015627.1 | 9 - 10 | 1568 | 65 | |
GenBank mRNA | BC118313.1 | - | 9 - 10 | 1568 | 65 | |
BT020922.1 | - | 9 - 10 | 1526 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |