Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015643.2 | NP_001015643.1 | 2 - 3 | 259 | 97 | |
GenBank mRNA | BC118160.1 | - | 2 - 3 | 259 | 97 | |
BT020996.1 | - | 2 - 3 | 345 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |