Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015643.2 | NP_001015643.1 | 6 - 7 | 821 | 131 | |
GenBank mRNA | BC118160.1 | - | 6 - 7 | 821 | 131 | |
BT020996.1 | - | 6 - 7 | 907 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |