Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017949.2 | NP_001017949.1 | 6 - 7 | 1424 | 65 | |
GenBank mRNA | BC118192.1 | - | 6 - 7 | 1424 | 65 | |
BT021692.1 | - | 6 - 7 | 949 | 65 | ||
MH037574.1 | - | 7 - 8 | 1123 | 65 | ||
MH428214.1 | - | 6 - 7 | 1296 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |