Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034232.2 | NP_001029404.1 | 3 - 4 | 351 | 91 | |
GenBank mRNA | BC113322.1 | - | 3 - 4 | 351 | 91 | |
BT021574.1 | - | 3 - 4 | 319 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |