Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TANGO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035328.2 | NP_001030405.2 | 4 - 5 | 417 | 76 | |
GenBank mRNA | BC113297.1 | - | 4 - 5 | 417 | 76 | |
BT021651.1 | - | 3 - 4 | 683 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |