Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOLA-DMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040481.3 | NP_001035571.1 | 3 - 4 | 838 | 141 | |
GenBank mRNA | BC118361.1 | - | 3 - 4 | 839 | 141 | |
BT021480.1 | - | 3 - 4 | 786 | 141 | ||
D76417.1 | - | 3 - 4 | 622 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |