Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045868.1 | NP_001039333.1 | 4 - 5 | 744 | 66 | |
GenBank mRNA | AJ420928.1 | - | 4 - 5 | 695 | 66 | |
BC105484.1 | - | 4 - 5 | 744 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |