Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045871.2 | NP_001039336.1 | 3 - 4 | 404 | 122 | |
GenBank mRNA | AF037273.1 | - | 2 - 3 | 146 | 122 | |
BC112857.1 | - | 3 - 4 | 405 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |