Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045899.1 | NP_001039364.1 | 5 - 6 | 984 | 73 | |
GenBank mRNA | BC105457.1 | - | 5 - 6 | 984 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |