Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045941.1 | NP_001039406.1 | 4 - 5 | 944 | 73 | |
GenBank mRNA | BC105455.1 | - | 4 - 5 | 944 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |