Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046006.1 | NP_001039471.1 | 7 - 8 | 1326 | 103 | |
GenBank mRNA | BC112499.1 | - | 7 - 8 | 1326 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |