Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046006.1 | NP_001039471.1 | 8 - 9 | 1434 | 94 | |
GenBank mRNA | BC112499.1 | - | 8 - 9 | 1434 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |