Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QRSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046336.1 | NP_001039801.1 | 9 - 10 | 1255 | 90 | |
GenBank mRNA | BC114078.1 | - | 9 - 10 | 1255 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |