Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046339.1 | NP_001039804.1 | 1 - 2 | 463 | 76 | |
GenBank mRNA | BC105471.1 | - | 1 - 2 | 463 | 76 | |
BC146029.1 | - | 1 - 2 | 666 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |