Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046379.1 | NP_001039844.1 | 3 - 4 | 315 | 81 | |
GenBank mRNA | BC113327.1 | - | 3 - 4 | 315 | 81 | |
BT030714.1 | - | 3 - 4 | 309 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |