Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046397.2 | NP_001039862.1 | 4 - 5 | 699 | 91 | |
GenBank mRNA | BC112681.1 | - | 4 - 5 | 700 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |