Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046507.2 | NP_001039972.1 | 5 - 6 | 788 | 97 | |
GenBank mRNA | BC112586.1 | - | 5 - 6 | 789 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |