Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046514.1 | NP_001039979.1 | 4 - 5 | 719 | 115 | |
GenBank mRNA | BC114150.1 | - | 4 - 5 | 719 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |