Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046528.1 | NP_001039993.1 | 2 - 3 | 201 | 80 | |
GenBank mRNA | BC113316.1 | - | 2 - 3 | 201 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |