Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMUB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046534.1 | NP_001039999.1 | 2 - 3 | 611 | 145 | |
GenBank mRNA | BC113228.1 | - | 2 - 3 | 611 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |