Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046537.2 | NP_001040002.1 | 5 - 6 | 626 | 152 | |
GenBank mRNA | BC114789.1 | - | 5 - 6 | 628 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |