Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046557.2 | NP_001040022.1 | 5 - 6 | 511 | 71 | |
GenBank mRNA | BC112500.1 | - | 5 - 6 | 512 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |