Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075127.2 | NP_001068595.1 | 3 - 4 | 493 | 99 | |
GenBank mRNA | AF047017.1 | - | 1 - 2 | 88 | 99 | |
BC118429.1 | - | 3 - 4 | 494 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |