Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075209.1 | NP_001068677.1 | 11 - 12 | 1034 | 72 | |
GenBank mRNA | BC116107.1 | - | 11 - 12 | 1034 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |